Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001286790.1 | NP_001273719.1 | 2 | 239 | |
NM_144571.2 | NP_653172.2 | 2 | 168 | ||
XM_011531806.2 | XP_011530108.1 | 3 | 279 | ||
XM_011531807.2 | XP_011530109.1 | 3 | 277 | ||
XM_011531808.2 | XP_011530110.1 | 2 | 225 | ||
XM_011531809.1 | XP_011530111.1 | 2 | 170 | ||
XM_011531810.1 | XP_011530112.1 | 2 | 730 | ||
XM_017007956.1 | XP_016863445.1 | 2 | 167 | ||
XM_017007957.1 | XP_016863446.1 | 2 | 517 | ||
GenBank mRNA | AK058188.1 | - | 2 | 146 | |
AK303417.1 | - | 2 | 217 | ||
AK304397.1 | - | 2 | 215 | ||
DA043791.1 | - | 2 | 168 | ||
DB454180.1 | - | 2 | 172 |