Gene Transcripts
- Gene Symbol
- BROX
- Entrez Gene ID
- 148362
- Gene Name
- BRO1 domain and CAAX motif containing
- Gene Aliases
- C1orf58
- Chromosome Location
- Chr. 1: 222712553 - 222735196 on Build GRCh38
- UniGene ID
- Hs.534965
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001288579.1 |
NP_001275508.1
|
5, 6 |
792 |
|
NM_001288580.1 |
NP_001275509.1
|
5, 6 |
498 |
|
NM_001288581.1 |
NP_001275510.1
|
6, 7 |
594 |
|
NM_144695.3 |
NP_653296.2
|
6, 7 |
888 |
|
XM_005273065.2 |
XP_005273122.2
|
6, 7 |
524 |
|
XM_005273069.4 |
XP_005273126.1
|
5, 6 |
760 |
|
XM_006711173.3 |
XP_006711236.1
|
7, 8 |
957 |
|
XM_011509212.2 |
XP_011507514.1
|
5, 6 |
851 |
|
XM_011509213.2 |
XP_011507515.1
|
5, 6 |
832 |
|
XM_011509214.2 |
XP_011507516.1
|
7, 8 |
757 |
|
XM_017000374.1 |
XP_016855863.1
|
8, 9 |
1082 |
|
XM_017000375.1 |
XP_016855864.1
|
4, 5 |
737 |
|
XM_017000376.1 |
XP_016855865.1
|
6, 7 |
885 |
|
GenBank mRNA |
AB276123.1 |
- |
5, 6 |
482 |
|
AK056983.1 |
- |
6, 7 |
877 |
|
AK293974.1 |
- |
5, 6 |
424 |
|
AK304445.1 |
- |
6, 7 |
539 |
|
AK315930.1 |
- |
5, 6 |
428 |
|
BC113635.1 |
- |
6, 7 |
625 |
8322687 |
BC113637.1 |
- |
6, 7 |
625 |
8322689 |
BC143811.1 |
- |
5, 6 |
529 |
9052326 |
View Full Product Details