Gene Transcripts
- Gene Symbol
- FBXL13
- Entrez Gene ID
- 222235
- Gene Name
- F-box and leucine rich repeat protein 13
- Gene Aliases
- DRC6, Fbl13
- Chromosome Location
- Chr. 7: 102699226 - 103074841 on Build GRCh38
- UniGene ID
- Hs.660029
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001111038.1 |
NP_001104508.1
|
10, 11 |
1136 |
|
NM_001287150.1 |
NP_001274079.1
|
10, 11 |
1136 |
|
NM_145032.3 |
NP_659469.3
|
10, 11 |
1313 |
|
NR_105043.1 |
|
11, 12 |
1227 |
|
XM_005250205.3 |
XP_005250262.1
|
11, 12 |
1219 |
|
XM_005250207.3 |
XP_005250264.1
|
11, 12 |
1219 |
|
XM_005250208.3 |
XP_005250265.1
|
11, 12 |
1219 |
|
XM_005250209.2 |
XP_005250266.1
|
10, 11 |
1117 |
|
XM_006715898.2 |
XP_006715961.1
|
5, 6 |
564 |
|
XM_011515928.2 |
XP_011514230.1
|
11, 12 |
1219 |
|
XM_011515929.2 |
XP_011514231.1
|
11, 12 |
1219 |
|
XM_011515930.2 |
XP_011514232.1
|
10, 11 |
1183 |
|
XM_011515932.2 |
XP_011514234.1
|
11, 12 |
1219 |
|
XM_017011850.1 |
XP_016867339.1
|
11, 12 |
1163 |
|
XM_017011851.1 |
XP_016867340.1
|
11, 12 |
1219 |
|
XM_017011852.1 |
XP_016867341.1
|
11, 12 |
1369 |
|
XM_017011853.1 |
XP_016867342.1
|
10, 11 |
1169 |
|
GenBank mRNA |
AK097537.1 |
- |
10, 11 |
1313 |
|
AL713709.1 |
- |
1, 2 |
35 |
|
AY359238.1 |
- |
10, 11 |
1020 |
|
AY359239.1 |
- |
10, 11 |
1020 |
|
BC020572.2 |
- |
4, 5 |
390 |
|
BC026121.1 |
- |
11, 12 |
1230 |
4826546 |
BC031285.1 |
- |
10, 11 |
1128 |
5298780 |
View Full Product Details