Gene Transcripts
- Gene Symbol
- SPACA7
- Entrez Gene ID
- 122258
- Gene Name
- sperm acrosome associated 7
- Gene Aliases
- C13orf28
- Chromosome Location
- Chr. 13: 112376300 - 112434695 on Build GRCh38
- UniGene ID
- Hs.97592
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK130866.1 |
- |
4 |
387 |
|
BC016750.2 |
- |
4 |
369 |
4837062 |
View Full Product Details