Gene Transcripts

Gene Symbol
OSR1
Entrez Gene ID
130497
Gene Name
odd-skipped related transciption factor 1
Gene Aliases
ODD
Chromosome Location
Chr. 2: 19351485 - 19358611 on Build GRCh38
UniGene ID
Hs.123933
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145260.2 NP_660303.1 3 1026
XM_006711942.3 XP_006712005.1 3 1895
GenBank mRNA AB082568.1 - 2 727
AK074591.1 - 3 1038
AK122756.1 - 2 3854
BC025712.1 - 3 1026 5227308

View Full Product Details