Gene Transcripts
- Gene Symbol
- TMEM99
- Entrez Gene ID
- 147184
- Gene Name
- transmembrane protein 99
- Gene Aliases
- Chromosome Location
- Chr. 17: 40819092 - 40836274 on Build GRCh38
- UniGene ID
- Hs.353163
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC015365.1 |
- |
3 |
574 |
|
View Full Product Details