Gene Transcripts
- Gene Symbol
- SHANK2-AS3
- Entrez Gene ID
- 220070
- Gene Name
- SHANK2 antisense RNA 3
- Gene Aliases
- C11orf76
- Chromosome Location
- Chr. 11: 70862790 - 70865115 on Build GRCh38
- UniGene ID
- Hs.326766
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_073536.1 |
|
2, 3 |
363 |
|
GenBank mRNA |
BC004224.2 |
- |
2, 3 |
363 |
|
View Full Product Details