Gene Transcripts

Gene Symbol
KIFC2
Entrez Gene ID
90990
Gene Name
kinesin family member C2
Gene Aliases
Chromosome Location
Chr. 8: 144466246 - 144474116 on Build GRCh38
UniGene ID
Hs.528713
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145754.3 NP_665697.1 10 1145
XM_005272357.3 XP_005272414.1 10 1235
XM_011517362.2 XP_011515664.1 10 1236
GenBank mRNA AK055045.1 - 7 523
BC017311.2 - 11 1127 5090622

View Full Product Details