Gene Transcripts

Gene Symbol
SLC39A13
Entrez Gene ID
91252
Gene Name
solute carrier family 39 member 13
Gene Aliases
LZT-Hs9
Chromosome Location
Chr. 11: 47407132 - 47416500 on Build GRCh38
UniGene ID
Hs.523664
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128225.2 NP_001121697.1 10 2339
NM_152264.4 NP_689477.2 10 2339
NR_134854.1 11 1576
XM_006718381.3 XP_006718444.1 10 2223
XM_006718383.3 XP_006718446.1 9 2115
XM_006718384.2 XP_006718447.1 10 2219
XM_006718385.2 XP_006718448.1 9 2111
XM_011520466.1 XP_011518768.1 10 2244
XM_011520467.1 XP_011518769.1 10 2573
XM_011520468.2 XP_011518770.1 10 2666
XM_011520469.1 XP_011518771.1 9 2136
XM_011520470.1 XP_011518772.1 9 2110
XM_017018540.1 XP_016874029.1 10 2197
XM_017018541.1 XP_016874030.1 9 2089
XM_017018542.1 XP_016874031.1 10 2193
GenBank mRNA AK098651.1 - 11 1576
AL133581.1 - 3 1312
BC008853.2 - 10 2206 4100291
BC019016.2 - 10 2204 4100057
BX647491.1 - 8 3821

View Full Product Details