Gene Transcripts
- Gene Symbol
- CHMP7
- Entrez Gene ID
- 91782
- Gene Name
- charged multivesicular body protein 7
- Gene Aliases
- Chromosome Location
- Chr. 8: 23243637 - 23262000 on Build GRCh38
- UniGene ID
- Hs.5019
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001317899.1 |
NP_001304828.1
|
2, 3 |
413 |
|
NM_152272.4 |
NP_689485.1
|
3, 4 |
1127 |
|
XM_006716415.2 |
XP_006716478.1
|
2, 3 |
583 |
|
XM_017013961.1 |
XP_016869450.1
|
2, 3 |
583 |
|
XM_017013962.1 |
XP_016869451.1
|
4, 5 |
1264 |
|
XM_017013963.1 |
XP_016869452.1
|
2, 3 |
390 |
|
XM_017013964.1 |
XP_016869453.1
|
3, 4 |
2276 |
|
XM_017013965.1 |
XP_016869454.1
|
2, 3 |
390 |
|
GenBank mRNA |
AK021776.1 |
- |
2, 3 |
557 |
|
AK092500.1 |
- |
2, 3 |
363 |
|
AK097202.1 |
- |
3, 4 |
1124 |
|
AK315664.1 |
- |
2, 3 |
564 |
|
AL833843.1 |
- |
2, 3 |
532 |
|
BC004344.1 |
- |
2, 3 |
355 |
|
BC019110.1 |
- |
2, 3 |
529 |
5093453 |
BC042050.1 |
- |
3, 4 |
1108 |
5551762 |
View Full Product Details