Gene Transcripts

Gene Symbol
ORAI3
Entrez Gene ID
93129
Gene Name
ORAI calcium release-activated calcium modulator 3
Gene Aliases
TMEM142C
Chromosome Location
Chr. 16: 30949084 - 30954938 on Build GRCh38
UniGene ID
Hs.745104
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152288.2 NP_689501.1 2 1054
GenBank mRNA BC006126.1 - 2 997
BC015555.1 - 2 1043 3878011
BC022786.1 - 2 997 3605904

View Full Product Details