Gene Transcripts

Gene Symbol
TMEM199
Entrez Gene ID
147007
Gene Name
transmembrane protein 199
Gene Aliases
C17orf32, CDG2P, VMA12, VPH2
Chromosome Location
Chr. 17: 28357581 - 28363683 on Build GRCh38
UniGene ID
Hs.594696
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152464.2 NP_689677.1 1, 2 307
GenBank mRNA AK223584.1 - 1, 2 241
AK291475.1 - 1, 2 230
AK311363.1 - 189
AY074907.1 - 1, 2 217
BC033113.1 - 1, 2 224 5429060
DC327155.1 - 1, 2 307

View Full Product Details