Gene Transcripts

Gene Symbol
LCA5L
Entrez Gene ID
150082
Gene Name
LCA5L, lebercilin like
Gene Aliases
C21orf13
Chromosome Location
Chr. 21: 39405844 - 39445796 on Build GRCh38
UniGene ID
Hs.517284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152505.3 NP_689718.1 4 427
XM_005260926.1 XP_005260983.1 4 385
XM_006723967.1 XP_006724030.1 3 430
XM_011529458.1 XP_011527760.1 5 836
XM_011529459.1 XP_011527761.1 5 495
XM_011529460.1 XP_011527762.1 6 635
XM_011529461.1 XP_011527763.1 6 742
XM_011529462.1 XP_011527764.1 4 385
XM_011529463.2 XP_011527765.1 4 438
XM_011529464.1 XP_011527766.1 5 464
XM_011529465.1 XP_011527767.1 4 383
XM_011529466.1 XP_011527768.1 4 621
XM_011529467.1 XP_011527769.1 5 699
XM_011529468.1 XP_011527770.1 4 341
XM_011529469.2 XP_011527771.1 4 728
XM_011529470.1 XP_011527772.1 3 224
XM_011529471.1 XP_011527773.1 3 430
GenBank mRNA BC031059.2 - 4 428
BC043006.1 - 4 644 5297703
BX647793.1 - 3 462
BX648744.1 - 1 55

View Full Product Details