Gene Transcripts

Gene Symbol
HORMAD2
Entrez Gene ID
150280
Gene Name
HORMA domain containing 2
Gene Aliases
CT46.2
Chromosome Location
Chr. 22: 30080069 - 30207425 on Build GRCh38
UniGene ID
Hs.120391
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011529917.2 XP_011528219.1 11 1723
GenBank mRNA BC030013.1 - 11 1474
BI829841.1 - 1 104

View Full Product Details