Gene Transcripts

Gene Symbol
STOX1
Entrez Gene ID
219736
Gene Name
storkhead box 1
Gene Aliases
C10orf24
Chromosome Location
Chr. 10: 68827537 - 68895942 on Build GRCh38
UniGene ID
Hs.37636
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130161.2 NP_001123633.1 3 1940
NM_152709.4 NP_689922.3 3 1940
XM_011539454.2 XP_011537756.1 3 1671
GenBank mRNA AK057891.1 - 1 330
AK124681.1 - 3 1635
AK127878.1 - 1 981
AY842014.1 - 3 1853
AY842017.1 - 3 1853
BC063627.1 - 1 321

View Full Product Details