Gene Transcripts

Gene Symbol
SLC44A5
Entrez Gene ID
204962
Gene Name
solute carrier family 44 member 5
Gene Aliases
CTL5
Chromosome Location
Chr. 1: 75202131 - 75724261 on Build GRCh38
UniGene ID
Hs.654821
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130058.1 NP_001123530.1 6, 7 398
NM_001320283.1 NP_001307212.1 4, 5 359
NM_001320285.1 NP_001307214.1 8, 9 558
NM_001320287.1 NP_001307216.1 7, 8 491
NM_152697.4 NP_689910.2 6, 7 398
XM_006710445.3 XP_006710508.1 4, 5 285
XM_011540986.2 XP_011539288.1 5, 6 376
XM_011540987.2 XP_011539289.1 7, 8 453
XM_017000607.1 XP_016856096.1 5, 6 424
XM_017000608.1 XP_016856097.1 5, 6 423
XM_017000609.1 XP_016856098.1 8, 9 1742
XM_017000610.1 XP_016856099.1 8, 9 820
XM_017000611.1 XP_016856100.1 6, 7 505
GenBank mRNA AK093170.1 - 6, 7 400
AK128063.1 - 7, 8 451
AK296924.1 - 5, 6 437
AK299573.1 - 4, 5 359
BC028743.2 - 6, 7 371 4828797
BC034580.1 - 8, 9 537 4824861
BC051740.1 - 8, 9 537 4824908
BG718149.1 - 6, 7 405
DB450780.1 - 6, 7 399
DC374918.1 - 4, 5 359

View Full Product Details