Gene Transcripts

Gene Symbol
BTBD9
Entrez Gene ID
114781
Gene Name
BTB domain containing 9
Gene Aliases
dJ322I12.1
Chromosome Location
Chr. 6: 38168451 - 38640426 on Build GRCh38
UniGene ID
Hs.603858
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099272.1 NP_001092742.1 3 762
NM_001172418.1 NP_001165889.1 2 547
NM_052893.1 NP_443125.1 4 962
NM_152733.2 NP_689946.2 2 499
XM_011514279.2 XP_011512581.1 5 1359
XM_011514280.2 XP_011512582.1 3 1040
XM_011514281.2 XP_011512583.1 3 1040
XM_011514282.2 XP_011512584.1 3 1040
XM_017010227.1 XP_016865716.1 4 1240
GenBank mRNA AB067467.1 - 3 628
AK057507.1 - 2 499
BC101354.2 - 2 420 40025568
BC101355.1 - 2 420 40025569
BC101356.2 - 2 420
BC101357.1 - 2 468 40025572
BX537534.1 - 3 495

View Full Product Details