Gene Transcripts
- Gene Symbol
- TCTEX1D1
- Entrez Gene ID
- 200132
- Gene Name
- Tctex1 domain containing 1
- Gene Aliases
- Chromosome Location
- Chr. 1: 66752147 - 66779051 on Build GRCh38
- UniGene ID
- Hs.479226
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098192.1 |
- |
5 |
1773 |
|
View Full Product Details