Gene Transcripts
- Gene Symbol
- NEDD1
- Entrez Gene ID
- 121441
- Gene Name
- neural precursor cell expressed, developmentally down-regulated 1
- Gene Aliases
- GCP-WD, TUBGCP7
- Chromosome Location
- Chr. 12: 96907222 - 96953691 on Build GRCh38
- UniGene ID
- Hs.270084
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135175.1 |
NP_001128647.1
|
3, 4 |
404 |
|
NM_001135176.1 |
NP_001128648.1
|
3, 4 |
323 |
|
NM_152905.3 |
NP_690869.1
|
4, 5 |
576 |
|
XM_005268644.2 |
XP_005268701.1
|
4, 5 |
310 |
|
XM_011537904.2 |
XP_011536206.1
|
3, 4 |
289 |
|
GenBank mRNA |
AK093221.1 |
- |
4, 5 |
514 |
|
AK290058.1 |
- |
4, 5 |
576 |
|
AK315821.1 |
- |
4, 5 |
542 |
|
AL832466.1 |
- |
3, 4 |
290 |
|
AL832468.1 |
- |
1, 2 |
249 |
|
BC020799.1 |
- |
4, 5 |
536 |
|
BC027605.2 |
- |
3, 4 |
404 |
4827191 |
View Full Product Details