Gene Transcripts
- Gene Symbol
- SPECC1
- Entrez Gene ID
- 92521
- Gene Name
- sperm antigen with calponin homology and coiled-coil domains 1
- Gene Aliases
- CYTSB, HCMOGT-1, HCMOGT1, NSP
- Chromosome Location
- Chr. 17: 20009301 - 20319026 on Build GRCh38
- UniGene ID
- Hs.431045
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033553.2 |
NP_001028725.1
|
4, 5 |
1922 |
|
NM_001033554.2 |
NP_001028726.1
|
2, 3 |
1831 |
|
NM_001033555.2 |
NP_001028727.1
|
2, 3 |
1831 |
|
NM_001243438.1 |
NP_001230367.1
|
2, 3 |
1831 |
|
NM_001243439.1 |
NP_001230368.1
|
4, 5 |
1981 |
|
NM_152904.4 |
NP_690868.3
|
4, 5 |
1922 |
|
XM_011524075.1 |
XP_011522377.1
|
3, 4 |
1874 |
|
XM_017025320.1 |
XP_016880809.1
|
4, 5 |
2016 |
|
XM_017025325.1 |
XP_016880814.1
|
4, 5 |
2016 |
|
GenBank mRNA |
AB041533.1 |
- |
4, 5 |
1922 |
|
AK295093.1 |
- |
4, 5 |
1954 |
|
AK295811.1 |
- |
1, 2 |
1458 |
|
AK298601.1 |
- |
1, 2 |
663 |
|
AK304186.1 |
- |
2, 3 |
1753 |
|
AK314341.1 |
- |
4, 5 |
1997 |
|
AY816326.1 |
- |
2, 3 |
1736 |
|
AY816327.1 |
- |
2, 3 |
1736 |
|
AY816328.1 |
- |
4, 5 |
1922 |
|
AY816329.1 |
- |
4, 5 |
1922 |
|
BC021123.1 |
- |
2, 3 |
1729 |
3614358 |
BC033618.1 |
- |
1, 2 |
946 |
|
BC050058.1 |
- |
2, 3 |
1666 |
5277450 |
View Full Product Details