Gene Transcripts

Gene Symbol
PPP1R21
Entrez Gene ID
129285
Gene Name
protein phosphatase 1 regulatory subunit 21
Gene Aliases
CCDC128, KLRAQ1
Chromosome Location
Chr. 2: 48440769 - 48515392 on Build GRCh38
UniGene ID
Hs.654619
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001135629.2 NP_001129101.1 5, 6 721
NM_001193475.1 NP_001180404.1 5, 6 721
NM_152994.4 NP_694539.1 5, 6 721
NR_024188.2 5, 6 760
XM_017003301.1 XP_016858790.1 5, 6 756
XM_017003302.1 XP_016858791.1 5, 6 756
GenBank mRNA AK122900.1 - 5, 6 731
AK172762.1 - 5, 6 693
AK307755.1 - 5, 6 644
AY092062.1 - 5, 6 672
AY134855.1 - 5, 6 721
BC040721.1 - 5, 6 622
BC111059.1 - 5, 6 663 5745902
BC117279.1 - 5, 6 633 40125830
BC143463.1 - 5, 6 633 9051971
BC143466.1 - 5, 6 633 9051974

View Full Product Details