Gene Transcripts
- Gene Symbol
- SPATA33
- Entrez Gene ID
- 124045
- Gene Name
- spermatogenesis associated 33
- Gene Aliases
- C16orf55
- Chromosome Location
- Chr. 16: 89657744 - 89670458 on Build GRCh38
- UniGene ID
- Hs.655171
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK056168.1 |
- |
3 |
1480 |
|
AK125673.1 |
- |
2 |
1419 |
|
AK128803.1 |
- |
2 |
1653 |
|
View Full Product Details