Gene Transcripts

Gene Symbol
PRICKLE1
Entrez Gene ID
144165
Gene Name
prickle planar cell polarity protein 1
Gene Aliases
EPM1B, RILP
Chromosome Location
Chr. 12: 42458338 - 42589770 on Build GRCh38
UniGene ID
Hs.524348
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001144881.1 NP_001138353.1 8 2621
NM_001144882.1 NP_001138354.1 8 2620
NM_001144883.1 NP_001138355.1 8 2617
NM_153026.2 NP_694571.2 8 2688
XM_011537946.1 XP_011536248.1 8 2439
XM_011537947.2 XP_011536249.1 8 2670
XM_017018838.1 XP_016874327.1 8 2484
XM_017018839.1 XP_016874328.1 8 2684
XM_017018840.1 XP_016874329.1 8 2504
GenBank mRNA AF399844.1 - 8 2594
AK056189.1 - 2 1212
AK056499.1 - 8 2594
AK122877.1 - 8 2622
AK122881.1 - 8 2562
BC114939.1 - 8 2529 40084889
BC114940.1 - 8 2530 40084893

View Full Product Details