Gene Transcripts
- Gene Symbol
- EML3
- Entrez Gene ID
- 256364
- Gene Name
- echinoderm microtubule associated protein like 3
- Gene Aliases
- ELP95
- Chromosome Location
- Chr. 11: 62602218 - 62613020 on Build GRCh38
- UniGene ID
- Hs.379785
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300793.1 |
NP_001287722.1
|
5, 6 |
956 |
|
NM_001300794.1 |
NP_001287723.1
|
5, 6 |
953 |
|
NM_153265.2 |
NP_694997.2
|
5, 6 |
953 |
|
XM_005273878.3 |
XP_005273935.1
|
5, 6 |
1211 |
|
XM_006718489.3 |
XP_006718552.1
|
5, 6 |
1328 |
|
XM_006718490.3 |
XP_006718553.1
|
5, 6 |
1328 |
|
XM_006718491.3 |
XP_006718554.1
|
5, 6 |
1325 |
|
XM_011544896.2 |
XP_011543198.1
|
5, 6 |
1325 |
|
XM_011544897.2 |
XP_011543199.1
|
5, 6 |
1328 |
|
XM_017017480.1 |
XP_016872969.1
|
5, 6 |
699 |
|
XM_017017481.1 |
XP_016872970.1
|
4, 5 |
585 |
|
XM_017017482.1 |
XP_016872971.1
|
5, 6 |
699 |
|
XM_017017483.1 |
XP_016872972.1
|
5, 6 |
756 |
|
GenBank mRNA |
AK093146.1 |
- |
4, 5 |
628 |
|
AK128679.1 |
- |
4, 5 |
1370 |
|
AK296901.1 |
- |
5, 6 |
718 |
|
AK310579.1 |
- |
7, 8 |
779 |
|
AL833388.1 |
- |
2, 3 |
208 |
|
BC075824.1 |
- |
4, 5 |
559 |
|
BC108280.1 |
- |
5, 6 |
737 |
3915493 |
View Full Product Details