Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153340.4 | NP_699171.3 | 10 | 1679 | |
XM_005270445.3 | XP_005270502.1 | 10 | 1810 | ||
XM_005270446.2 | XP_005270503.1 | 10 | 1806 | ||
XM_011540638.2 | XP_011538940.1 | 9 | 1709 | ||
XM_011540639.2 | XP_011538941.1 | 10 | 3079 | ||
XM_011540641.2 | XP_011538943.1 | 10 | 2412 | ||
XM_011540644.2 | XP_011538946.1 | 12 | 1894 | ||
XM_011540645.1 | XP_011538947.1 | 9 | 1655 | ||
XM_017000267.1 | XP_016855756.1 | 11 | 2541 | ||
XM_017000268.1 | XP_016855757.1 | 11 | 1874 | ||
XM_017000269.1 | XP_016855758.1 | 11 | 2541 | ||
XM_017000270.1 | XP_016855759.1 | 11 | 1942 | ||
GenBank mRNA | AK090460.1 | - | 7 | 3357 |