Gene Transcripts
- Gene Symbol
- VRK1
- Entrez Gene ID
- 7443
- Gene Name
- vaccinia related kinase 1
- Gene Aliases
- PCH1, PCH1A
- Chromosome Location
- Chr. 14: 96797309 - 96881614 on Build GRCh38
- UniGene ID
- Hs.422662
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003384.2 |
NP_003375.1
|
10, 11 |
1003 |
|
XM_006720247.3 |
XP_006720310.1
|
10, 11 |
1041 |
|
XM_011537132.1 |
XP_011535434.1
|
9, 10 |
902 |
|
XM_017021622.1 |
XP_016877111.1
|
9, 10 |
902 |
|
XM_017021624.1 |
XP_016877113.1
|
10, 11 |
1041 |
|
XM_017021625.1 |
XP_016877114.1
|
10, 11 |
1082 |
|
XM_017021626.1 |
XP_016877115.1
|
9, 10 |
920 |
|
GenBank mRNA |
AB000449.1 |
- |
10, 11 |
972 |
|
AK290110.1 |
- |
10, 11 |
991 |
|
BC103761.1 |
- |
10, 11 |
981 |
5773877 |
BC112075.1 |
- |
10, 11 |
971 |
8327543 |
BC113510.1 |
- |
10, 11 |
971 |
8322562 |
View Full Product Details