Gene Transcripts
- Gene Symbol
- HSCB
- Entrez Gene ID
- 150274
- Gene Name
- HscB mitochondrial iron-sulfur cluster cochaperone
- Gene Aliases
- DNAJC20, HSC20, JAC1
- Chromosome Location
- Chr. 22: 28741992 - 28757518 on Build GRCh38
- UniGene ID
- Hs.632780
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_172002.4 |
NP_741999.3
|
3, 4 |
484 |
|
NR_134560.1 |
|
3, 4 |
429 |
|
XM_005261359.3 |
XP_005261416.1
|
3, 4 |
485 |
|
XM_017028620.1 |
XP_016884109.1
|
3, 4 |
288 |
|
GenBank mRNA |
AY191719.1 |
- |
3, 4 |
434 |
|
BC000004.1 |
- |
3, 4 |
493 |
|
BC065569.1 |
- |
3, 4 |
457 |
6171959 |
BG696148.1 |
- |
3, 4 |
418 |
|
CR456462.1 |
- |
3, 4 |
452 |
|
DW414316.1 |
- |
3, 4 |
249 |
|
View Full Product Details