Gene Transcripts

Gene Symbol
POC1B
Entrez Gene ID
282809
Gene Name
POC1 centriolar protein B
Gene Aliases
CORD20, PIX1, TUWD12, WDR51B
Chromosome Location
Chr. 12: 89419718 - 89526262 on Build GRCh38
UniGene ID
Hs.25130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199777.1 NP_001186706.1 5 969
NM_172240.2 NP_758440.1 6 988
NR_037659.1 5 816
NR_037660.1 4 797
GenBank mRNA AB053266.1 - 5 478
AK027829.1 - 2 187
AK074772.1 - 6 726
AK123106.1 - 5 966
AK295596.1 - 5 554
AL832918.1 - 4 453
BC026080.2 - 6 750 4827689
BU521431.1 - 4 466
EF011619.1 - 6 750

View Full Product Details