Gene Transcripts

Gene Symbol
DAOA
Entrez Gene ID
267012
Gene Name
D-amino acid oxidase activator
Gene Aliases
LG72, SG72
Chromosome Location
Chr. 13: 105465867 - 105491034 on Build GRCh38
UniGene ID
Hs.381382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161812.1 NP_001155284.1 3 278
NM_001161814.1 NP_001155286.1 4 472
NM_172370.4 NP_758958.3 3 232
XM_005254042.1 XP_005254099.1 3 430
GenBank mRNA AY138546.1 - 3 205
AY138547.1 - 4 472
AY170469.2 - 4 294
AY170470.2 - 4 294
AY170471.1 - 3 172
AY223901.1 - 3 430
BC121091.1 - 3 200 40119921
DQ343761.1 - 3 479
DQ357223.1 - 3 224
DQ386869.1 - 3 178
DQ386870.1 - 3 224

View Full Product Details