Gene Transcripts

Gene Symbol
CYP4F22
Entrez Gene ID
126410
Gene Name
cytochrome P450 family 4 subfamily F member 22
Gene Aliases
ARCI5, INLNE, LI3
Chromosome Location
Chr. 19: 15508487 - 15552317 on Build GRCh38
UniGene ID
Hs.156452
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173483.3 NP_775754.2 4 436
XM_011527692.2 XP_011525994.1 5 678
XM_011527693.2 XP_011525995.1 4 448
GenBank mRNA AK096820.1 - 4 436
BC069351.1 - 3 373 7262372
BC093894.1 - 3 310 7939739
BC093896.1 - 3 310 7939741
DA874258.1 - 4 436

View Full Product Details