Gene Transcripts
- Gene Symbol
- C14orf80
- Entrez Gene ID
- 283643
- Gene Name
- chromosome 14 open reading frame 80
- Gene Aliases
- Chromosome Location
- Chr. 14: 105489855 - 105499248 on Build GRCh38
- UniGene ID
- Hs.720306
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC016028.1 |
- |
7 |
1279 |
|
View Full Product Details