Gene Transcripts

Gene Symbol
ARL10
Entrez Gene ID
285598
Gene Name
ADP ribosylation factor like GTPase 10
Gene Aliases
ARL10A
Chromosome Location
Chr. 5: 176365468 - 176415085 on Build GRCh38
UniGene ID
Hs.424552
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173664.5 NP_775935.1 4 1381
XM_011534531.2 XP_011532833.1 4 875

View Full Product Details