Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001206955.1 | NP_001193884.1 | 19 | 2586 | |
NM_001206956.1 | NP_001193885.1 | 11 | 1305 | ||
NM_175607.2 | NP_783200.1 | 20 | 2541 | ||
NM_175613.2 | NP_783302.1 | 11 | 1308 | ||
XM_006713004.3 | XP_006713067.1 | 19 | 2551 | ||
XM_011533425.2 | XP_011531727.1 | 19 | 2554 | ||
XM_011533426.2 | XP_011531728.1 | 20 | 2773 | ||
XM_011533427.2 | XP_011531729.1 | 20 | 2754 | ||
XM_011533428.2 | XP_011531730.1 | 18 | 2743 | ||
XM_011533429.2 | XP_011531731.1 | 18 | 2678 | ||
XM_011533430.2 | XP_011531732.1 | 18 | 2639 | ||
XM_011533431.2 | XP_011531733.1 | 17 | 5388 | ||
XM_017005782.1 | XP_016861271.1 | 20 | 2496 | ||
XM_017005783.1 | XP_016861272.1 | 21 | 3370 | ||
XM_017005784.1 | XP_016861273.1 | 19 | 2831 | ||
XM_017005785.1 | XP_016861274.1 | 20 | 2478 | ||
XM_017005786.1 | XP_016861275.1 | 18 | 2740 | ||
XM_017005787.1 | XP_016861276.1 | 18 | 2636 | ||
GenBank mRNA | AF549455.1 | - | 17 | 2584 |