Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000561.3 | NP_000552.2 | 5 | 389 | |
NM_146421.2 | NP_666533.1 | 5 | 389 | ||
XM_005270782.4 | XP_005270839.1 | 5 | 407 | ||
GenBank mRNA | AY510272.1 | - | 5 | 311 | |
AY532926.1 | - | 5 | 311 | ||
AY532927.1 | - | 5 | 311 | ||
BC024005.2 | - | 5 | 345 | 4823313 | |
BC036805.1 | - | 5 | 326 | ||
BQ436476.1 | - | 5 | 375 | ||
CR541868.1 | - | 5 | 311 | ||
J03817.1 | - | 5 | 326 | ||
KJ534856.1 | - | 5 | 311 | ||
KJ534857.1 | - | 5 | 311 | ||
KJ534858.1 | - | 5 | 311 | ||
KJ534998.1 | - | 4 | 229 | ||
KJ535021.1 | - | 4 | 362 | ||
X08020.1 | - | 5 | 335 |