Gene Transcripts

Gene Symbol
HIST1H4E
Entrez Gene ID
8367
Gene Name
histone cluster 1, H4e
Gene Aliases
H4/j, H4FJ
Chromosome Location
Chr. 6: 26204645 - 26205021 on Build GRCh38
UniGene ID
Hs.662174
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003545.3 NP_003536.1 1 342
GenBank mRNA BC012587.1 - 1 381
BC050615.1 - 1 358

View Full Product Details