Gene Transcripts

Gene Symbol
VCX
Entrez Gene ID
26609
Gene Name
variable charge, X-linked
Gene Aliases
VCX-10r, VCX-B1, VCX1, VCX10R, VCXB1
Chromosome Location
Chr. X: 7842262 - 7844143 on Build GRCh38
UniGene ID
Hs.278906
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013452.2 NP_038480.2 3 841
XM_011545490.2 XP_011543792.1 3 673
GenBank mRNA AF159129.1 - 2 648
AF167081.2 - 3 841

Gene Symbol
VCX3A
Entrez Gene ID
51481
Gene Name
variable charge, X-linked 3A
Gene Aliases
VCX-8r, VCX-A, VCX3, VCX8R, VCXA
Chromosome Location
Chr. X: 6533618 - 6535118 on Build GRCh38
UniGene ID
Hs.170076
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC126902.1 - 4 678

Gene Symbol
VCX3B
Entrez Gene ID
425054
Gene Name
variable charge, X-linked 3B
Gene Aliases
VCX-C, VCXC
Chromosome Location
Chr. X: 8464830 - 8466510 on Build GRCh38
UniGene ID
Hs.279737
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001888.3 NP_001001888.3 3 1049
GenBank mRNA BC098143.1 - 4 671

View Full Product Details