Gene Transcripts

Gene Symbol
IFITM3
Entrez Gene ID
10410
Gene Name
interferon induced transmembrane protein 3
Gene Aliases
1-8U, DSPA2b, IP15
Chromosome Location
Chr. 11: 319673 - 320914 on Build GRCh38
UniGene ID
Hs.374650
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021034.2 NP_066362.2 2 657
NR_049759.1 2 652
GenBank mRNA X57352.1 - 2 793

View Full Product Details