Gene Transcripts

Gene Symbol
SCRT1
Entrez Gene ID
83482
Gene Name
scratch family transcriptional repressor 1
Gene Aliases
SCRT, ZNF898
Chromosome Location
Chr. 8: 144330565 - 144336281 on Build GRCh38
UniGene ID
Hs.31746
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031309.5 NP_112599.2 2 1432
GenBank mRNA BC014675.1 - 1 769

View Full Product Details