Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001306146.1 | NP_001293075.1 | 6 | 947 | |
NM_002112.3 | NP_002103.2 | 6 | 947 | ||
XM_017022094.1 | XP_016877583.1 | 6 | 711 | ||
XM_017022095.1 | XP_016877584.1 | 6 | 711 | ||
XM_017022096.1 | XP_016877585.1 | 5 | 496 | ||
XM_017022097.1 | XP_016877586.1 | 4 | 794 | ||
XM_017022098.1 | XP_016877587.1 | 3 | 314 | ||
XM_017022099.1 | XP_016877588.1 | 6 | 711 | ||
GenBank mRNA | AK308833.1 | - | 6 | 947 | |
AK310479.1 | - | 4 | 473 | ||
AK311072.1 | - | 6 | 666 | ||
BC130527.1 | - | 6 | 648 | 40146558 | |
BC144173.1 | - | 6 | 648 | 9052693 | |
M60445.1 | - | 6 | 656 | ||
X54297.1 | - | 6 | 707 |