Gene Transcripts

Gene Symbol
GLUD2
Entrez Gene ID
2747
Gene Name
glutamate dehydrogenase 2
Gene Aliases
GDH2, GLUDP1
Chromosome Location
Chr. X: 121047608 - 121049942 on Build GRCh38
UniGene ID
Hs.368538
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012084.3 NP_036216.2 1 337
GenBank mRNA AK313356.1 - 1 334
BC050732.2 - 1 337 6164919

View Full Product Details