Gene Transcripts
- Gene Symbol
- LOC107985476
- Entrez Gene ID
- 107985476
- Gene Name
- putative uncharacterized protein encoded by LINC00205
- Gene Aliases
- Chromosome Location
- Chr. 21: 45293658 - 45294696 on Build GRCh38
- Species
- Homo sapiens
- Gene Symbol
- LINC00205
- Entrez Gene ID
- 102723489
- Gene Name
- long intergenic non-protein coding RNA 205
- Gene Aliases
- C21orf86, NCRNA00205
- Chromosome Location
- Chr. 21: 45290197 - 45296026 on Build GRCh38
- UniGene ID
- Hs.11637
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF426264.1 |
- |
2 |
800 |
|
View Full Product Details