Gene Transcripts

Gene Symbol
AQP11
Entrez Gene ID
282679
Gene Name
aquaporin 11
Gene Aliases
AQPX1
Chromosome Location
Chr. 11: 77589635 - 77610356 on Build GRCh38
UniGene ID
Hs.503345
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173039.2 NP_766627.1 2 1057
GenBank mRNA AB028147.1 - 2 1004
AK075346.1 - 2 699
AK311096.1 - 3 454
BC040443.1 - 2 1057 5728157

View Full Product Details