Gene Transcripts

Gene Symbol
TOR1B
Entrez Gene ID
27348
Gene Name
torsin family 1 member B
Gene Aliases
DQ1
Chromosome Location
Chr. 9: 129803143 - 129811424 on Build GRCh38
UniGene ID
Hs.252682
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317893.1 NP_001304822.1 2 297
NM_001317894.1 NP_001304823.1 2 297
NM_014506.2 NP_055321.1 2 297
NR_133948.1 2 297
NR_134113.1 2 297
XM_005251927.3 XP_005251984.1 2 292
GenBank mRNA AF007872.1 - 1 18
AF317129.1 - 2 287
BC015578.2 - 2 242 4589479
BE315222.1 - 2 171
BP366330.1 - 2 297

View Full Product Details