Gene Transcripts

Gene Symbol
HUWE1
Entrez Gene ID
10075
Gene Name
HECT, UBA and WWE domain containing 1, E3 ubiquitin protein ligase
Gene Aliases
ARF-BP1, HECTH9, HSPC272, Ib772, LASU1, MULE, URE-B1, UREB1
Chromosome Location
Chr. X: 53532096 - 53686729 on Build GRCh38
UniGene ID
Hs.136905
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031407.6 NP_113584.3 29 3532
XM_005261965.3 XP_005262022.1 28 3599
XM_011530747.1 XP_011529049.1 30 3775
XM_011530751.2 XP_011529053.1 29 3643
XM_017029191.1 XP_016884680.1 28 6527
XM_017029192.1 XP_016884681.1 28 6527
XM_017029193.1 XP_016884682.1 28 6506
XM_017029194.1 XP_016884683.1 28 6527
XM_017029195.1 XP_016884684.1 28 6527
XM_017029196.1 XP_016884685.1 28 6527
XM_017029197.1 XP_016884686.1 28 6527
XM_017029198.1 XP_016884687.1 29 3869
XM_017029199.1 XP_016884688.1 30 3796
XM_017029200.1 XP_016884689.1 29 3666
XM_017029201.1 XP_016884690.1 29 3683
XM_017029202.1 XP_016884691.1 29 3659
XM_017029203.1 XP_016884692.1 29 3665
XM_017029204.1 XP_016884693.1 27 6277
XM_017029205.1 XP_016884694.1 27 6257
XM_017029206.1 XP_016884695.1 28 6527
GenBank mRNA AB071605.1 - 2 231
AK056671.1 - 12 1758
AK091352.1 - 1 50
AK294575.1 - 16 2132
AY772009.1 - 27 3177
AY929612.1 - 26 3129
DQ097177.1 - 29 3531

View Full Product Details