Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001040022.1 | NP_001035111.1 | 8 | 1613 | |
NM_001040023.1 | NP_001035112.1 | 7 | 1521 | ||
NM_080792.2 | NP_542970.1 | 8 | 1280 | ||
XM_005260669.3 | XP_005260726.1 | 8 | 1663 | ||
XM_005260670.3 | XP_005260727.1 | 8 | 1279 | ||
XM_006723545.3 | XP_006723608.1 | 5 | 850 | ||
XM_011529173.2 | XP_011527475.1 | 7 | 1268 | ||
GenBank mRNA | AB023430.1 | - | 8 | 1280 | |
AK290776.1 | - | 8 | 1386 | ||
AK298243.1 | - | 8 | 1231 | ||
AK312521.1 | - | 7 | 1334 | ||
BC026692.1 | - | 8 | 1285 | 3916836 | |
BC033092.1 | - | 7 | 1273 | 4589298 | |
BC038510.2 | - | 7 | 1518 | 5261789 | |
BC075849.1 | - | 8 | 1459 | 4589732 | |
D86043.1 | - | 7 | 1252 | ||
Y10375.1 | - | 7 | 1262 | ||
Y11047.1 | - | 6 | 1028 |