Gene Transcripts

Gene Symbol
RHOBTB1
Entrez Gene ID
9886
Gene Name
Rho related BTB domain containing 1
Gene Aliases
Chromosome Location
Chr. 10: 60869438 - 61001486 on Build GRCh38
UniGene ID
Hs.737374
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242359.1 NP_001229288.1 5 672
NM_014836.4 NP_055651.1 4 600
NR_024554.2 5 474
NR_024555.1 5 438
XM_006718084.1 XP_006718147.1 4 408
XM_006718085.1 XP_006718148.1 4 435
XM_011540424.1 XP_011538726.1 5 466
XM_011540425.1 XP_011538727.1 5 466
XM_011540426.1 XP_011538728.1 5 466
XM_011540428.1 XP_011538730.1 5 466
XM_017017000.1 XP_016872489.1 4 655
XM_017017001.1 XP_016872490.1 4 651
XM_017017002.1 XP_016872491.1 4 651
XM_017017003.1 XP_016872492.1 5 484
XM_017017004.1 XP_016872493.1 5 486
XM_017017005.1 XP_016872494.1 4 391
XM_017017006.1 XP_016872495.1 4 440
XM_017017008.1 XP_016872497.1 4 651
GenBank mRNA AB018283.3 - 4 491
AK075129.1 - 5 438
AK307630.1 - 4 356
BC032848.1 - 5 649 4830471
BC041791.1 - 4 391 5297756
BC093035.1 - 6 510
CB959762.1 - 462
CF593541.1 - 4 415
CN286750.1 - 5 427
DA781491.1 - 5 466
DA859805.1 - 4 435
DR004641.1 - 5 481

View Full Product Details