Gene Transcripts

Gene Symbol
CXXC5
Entrez Gene ID
51523
Gene Name
CXXC finger protein 5
Gene Aliases
CF5, HSPC195, RINF, WID
Chromosome Location
Chr. 5: 139647299 - 139683885 on Build GRCh38
UniGene ID
Hs.189119
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317199.1 NP_001304128.1 3 1317
NM_001317200.1 NP_001304129.1 2 754
NM_001317201.1 NP_001304130.1 2 806
NM_001317202.1 NP_001304131.1 2 770
NM_001317203.1 NP_001304132.1 3 978
NM_001317204.1 NP_001304133.1 3 848
NM_001317205.1 NP_001304134.1 2 847
NM_001317206.1 NP_001304135.1 4 1074
NM_001317207.1 NP_001304136.1 2 756
NM_001317208.1 NP_001304137.1 3 862
NM_001317209.1 NP_001304138.1 2 768
NM_001317210.1 NP_001304139.1 3 856
NM_001317211.1 NP_001304140.1 3 850
NM_016463.8 NP_057547.5 2 1223
XM_017009573.1 XP_016865062.1 2 1093
GenBank mRNA AB097005.1 - 2 800
AB097032.1 - 2 800
AF151029.1 - 1 429
AK001782.1 - 2 745
AK024338.1 - 2 1223
AK314420.1 - 1 507
AY007103.1 - 1 868
BC002490.2 - 1 559 3050465
BC006428.1 - 2 1154 3343683
BC017439.1 - 2 1115 2989328
BC024040.1 - 2 1144 3925666
BC041013.1 - 2 1144 4139961
BC050046.1 - 2 1116
GQ379202.1 - 1 509

View Full Product Details