Gene Transcripts

Gene Symbol
FAM84B
Entrez Gene ID
157638
Gene Name
family with sequence similarity 84 member B
Gene Aliases
BCMP101, NSE2
Chromosome Location
Chr. 8: 126552438 - 126558499 on Build GRCh38
UniGene ID
Hs.741352
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174911.4 NP_777571.1 2 1281
XM_017013107.1 XP_016868596.1 2 1328
XM_017013108.1 XP_016868597.1 3 1260
GenBank mRNA AJ417849.1 - 2 1208
AK292824.1 - 2 1206
BC052957.1 - 2 1036 5262890

View Full Product Details