Gene Transcripts

Gene Symbol
ARMC5
Entrez Gene ID
79798
Gene Name
armadillo repeat containing 5
Gene Aliases
AIMAH2
Chromosome Location
Chr. 16: 31458273 - 31467167 on Build GRCh38
UniGene ID
Hs.732945
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024742.2 NP_079018.1 4 2624
GenBank mRNA AK024430.1 - 4 2117
AY217348.1 - 4 2624

View Full Product Details