Gene Transcripts

Gene Symbol
SHISA7
Entrez Gene ID
729956
Gene Name
shisa family member 7
Gene Aliases
Chromosome Location
Chr. 19: 55428738 - 55442863 on Build GRCh38
UniGene ID
Hs.6664
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145176.1 NP_001138648.1 4 3627
GenBank mRNA AK094959.1 - 1 246
AK127223.1 - 1 1798

View Full Product Details